Assay Details
Gene Symbol: FUBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006717313.2 | XP_006717376.1 | 2 - 3 | 1045 | 112 | |
XM_017015269.1 | XP_016870758.1 | 2 - 3 | 1045 | 112 | ||
XM_017015270.1 | XP_016870759.1 | 2 - 3 | 1045 | 112 | ||
XM_017015273.1 | XP_016870762.1 | 2 - 3 | 1045 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006717313.2, XM_017015269.1, XM_017015270.1, XM_017015273.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of gene expression
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
poly(A) RNA binding
transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of gene expression
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
