Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057308.hg.1 |
TC01003733.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057308.hg.1 |
ENST00000367187.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057308.hg.1 |
ENST00000424712.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057308.hg.1 |
NM_002646.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229044.hg.1 |
ENST00000367187.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229044.hg.1 |
ENST00000424712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229044.hg.1 |
NM_002646.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229044.hg.1 |
ENST00000367187.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229044.hg.1 |
ENST00000424712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229044.hg.1 |
NM_002646.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229044.hg.1 |
TC0100017029.hg.1 |
Clariom D TC |