Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010325.hg.1 |
TC01000624.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010325.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010325.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010325.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010328.hg.1 |
TC01000624.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010328.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010328.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010328.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015220.hg.1 |
ENST00000392337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015220.hg.1 |
ENST00000392337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010329.hg.1 |
TC01000624.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010329.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010329.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159000.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159000.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159000.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159000.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159000.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159000.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159000.hg.1 |
TC0100008271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100158998.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158998.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158998.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158998.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158998.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158998.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158998.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158998.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158998.hg.1 |
TC0100008271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100158999.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158999.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158999.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158999.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158999.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158999.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158999.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158999.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158999.hg.1 |
TC0100008271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100159001.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159001.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159001.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159001.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159001.hg.1 |
TC0100008271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015220.hg.1 |
ENST00000392337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015220.hg.1 |
ENST00000392337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000242719.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000486691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
ENST00000494873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008271.hg.1 |
NM_014372.4 |
Refseq |