Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
TC01002652.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
ENST00000352171.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
ENST00000354831.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
ENST00000371665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
ENST00000475715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
ENST00000485608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
ENST00000539524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
ENST00000544028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
ENST00000546169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
NM_001101662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
NM_001242361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041535.hg.1 |
NM_002525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000352171.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000354831.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000475715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000485608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000539524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000544028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000615102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
NM_001101662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
NM_001242361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
NM_002525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000352171.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000354831.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000475715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000485608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000539524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000544028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
ENST00000615102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
NM_001101662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
NM_001242361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
NM_002525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206056.hg.1 |
TC0100014188.hg.1 |
Clariom D TC |