Assay Details
| Gene Symbol: TECTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005422.2 | NP_005413.2 | 14 - 15 | 4976 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001105.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000478058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000478058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | ENST00000478058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014088.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014088.hg.1 | ENST00000392793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014088.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014088.hg.1 | TC11001105.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100169778.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100169778.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100169778.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100169778.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100169778.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100169778.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100169778.hg.1 | TC1100009302.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000264037.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000392793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000478058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000478058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | ENST00000478058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009302.hg.1 | NM_005422.2 | Refseq |

TECTA
Genomic Map