Assay Details
Gene Symbol: ARCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142281.1 | NP_001135753.1 | 3 - 4 | 549 | 112 | |
NM_001655.4 | NP_001646.2 | 4 - 5 | 813 | 112 | ||
XM_005271542.3 | XP_005271599.1 | 4 - 5 | 837 | 112 | ||
GenBank mRNA | AK304019.1 | - | 3 - 4 | 546 | 112 | |
AK312298.1 | - | 4 - 5 | 791 | 112 | ||
BC093636.1 | - | 4 - 5 | 740 | 7939481 | 112 | |
BC093638.1 | - | 4 - 5 | 737 | 7939483 | 112 | |
BX640860.1 | - | 5 - 6 | 941 | 112 | ||
X81197.1 | - | 4 - 5 | 769 | 112 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001066.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11013464.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11013464.hg.1 | ENST00000359415.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11013464.hg.1 | ENST00000392859.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11013464.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11013464.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11013464.hg.1 | TC11001066.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100168654.hg.1 | TC1100009210.hg.1 | Clariom D TC |
