Assay Details
Gene Symbol: ARCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142281.1 | NP_001135753.1 | 7 - 8 | 1137 | 157 | |
NM_001655.4 | NP_001646.2 | 8 - 9 | 1401 | 157 | ||
GenBank mRNA | AK312298.1 | - | 8 - 9 | 1379 | 157 | |
BC093636.1 | - | 8 - 9 | 1328 | 7939481 | 157 | |
BC093638.1 | - | 8 - 9 | 1325 | 7939483 | 157 | |
BX640860.1 | - | 9 - 10 | 1529 | 157 | ||
X81197.1 | - | 8 - 9 | 1357 | 157 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001066.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11013468.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11013468.hg.1 | ENST00000359415.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11013468.hg.1 | ENST00000392859.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11013468.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11013468.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11013468.hg.1 | TC11001066.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100168658.hg.1 | TC1100009210.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000264028.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000359415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000392859.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000534182.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | ENST00000614498.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001142281.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009210.hg.1 | NM_001655.4 | Refseq |
