Assay Details
| Gene Symbol: DDX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257191.2 | NP_001244120.1 | 8 - 9 | 1219 | 91 | |
| NM_004397.5 | NP_004388.2 | 8 - 9 | 1291 | 91 | ||
| XM_005271417.3 | XP_005271474.1 | 8 - 9 | 1236 | 91 | ||
| XM_011542645.1 | XP_011540947.1 | 7 - 8 | 1144 | 91 | ||
| GenBank mRNA | AK313244.1 | - | 8 - 9 | 1219 | 91 | |
| BC065007.1 | - | 7 - 8 | 1150 | 6163439 | 91 | |
| D17532.1 | - | 8 - 9 | 1164 | 91 | ||
| Z11685.1 | - | 8 - 9 | 1191 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002353.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translation
RNA secondary structure unwinding
viral RNA genome packaging
stem cell population maintenance
cytoplasmic mRNA processing body assembly
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
negative regulation of neuron differentiation
cell-cell adhesion
RNA helicase activity
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding
protein domain specific binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
regulation of translation
RNA secondary structure unwinding
viral RNA genome packaging
stem cell population maintenance
cytoplasmic mRNA processing body assembly
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
negative regulation of neuron differentiation
cell-cell adhesion
RNA helicase activity
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding
protein domain specific binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000531971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000531971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000531971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000531971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000533239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000533239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000533239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000533239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |

DDX6
Genomic Map