Assay Details
| Gene Symbol: RGSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017001194.1 | XP_016856683.1 | 5 - 6 | 475 | 126 | |
| GenBank mRNA | DB097250.1 | - | 5 - 6 | 475 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017001194.1 transcripts.
Set Membership: |
|

RGSL1
Genomic Map