Assay Details
| Gene Symbol: USP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243759.1 | NP_001230688.1 | 6 - 7 | 723 | 88 | |
| NM_004205.4 | NP_004196.4 | 7 - 8 | 1538 | 88 | ||
| NM_171997.2 | NP_741994.1 | 6 - 7 | 803 | 88 | ||
| XM_005271721.4 | XP_005271778.1 | 7 - 8 | 1411 | 88 | ||
| XM_005271722.2 | XP_005271779.1 | 7 - 8 | 1417 | 88 | ||
| XM_017018539.1 | XP_016874028.1 | 6 - 7 | 499 | 88 | ||
| GenBank mRNA | AF079564.1 | - | 6 - 7 | 541 | 88 | |
| AF440755.1 | - | 6 - 7 | 803 | 88 | ||
| AK292255.1 | - | 6 - 7 | 726 | 88 | ||
| AK301805.1 | - | 6 - 7 | 608 | 88 | ||
| BC002854.1 | - | 7 - 8 | 1419 | 3635143 | 88 | |
| BC002955.1 | - | 7 - 8 | 1419 | 3543435 | 88 | |
| BC041366.2 | - | 6 - 7 | 708 | 5273400 | 88 | |
More Information
Set Membership: |
|

USP2
Genomic Map