Assay Details
Gene Symbol: TPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012112.4 | NP_036244.2 | 1 - 2 | 520 | 119 | |
GenBank mRNA | AF098158.1 | - | 1 - 2 | 313 | 119 | |
AF287265.1 | - | 1 - 2 | 286 | 119 | ||
AK312549.1 | - | 1 - 2 | 298 | 119 | ||
BC004136.1 | - | 1 - 2 | 250 | 3509275 | 119 | |
BC020207.1 | - | 1 - 2 | 279 | 5015279 | 119 | |
BU181208.1 | - | 1 - 2 | 362 | 119 | ||
CB960160.1 | - | 376 | 119 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000198.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF098158.1,AF287265.1,AK312549.1,BC004136.1,BC020207.1,NM_012112.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
apoptotic process
mitotic nuclear division
cell proliferation
activation of protein kinase activity
cell division
regulation of mitotic spindle organization
mitotic spindle assembly
regulation of signal transduction by p53 class mediator
protein binding
ATP binding
GTP binding
protein kinase binding
importin-alpha family protein binding
G2/M transition of mitotic cell cycle
apoptotic process
mitotic nuclear division
cell proliferation
activation of protein kinase activity
cell division
regulation of mitotic spindle organization
mitotic spindle assembly
regulation of signal transduction by p53 class mediator
protein binding
ATP binding
GTP binding
protein kinase binding
importin-alpha family protein binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20002361.hg.1 | ENST00000300403.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002361.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002361.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002361.hg.1 | TC20000198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20002362.hg.1 | ENST00000300403.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002362.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002362.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002362.hg.1 | TC20000198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20002366.hg.1 | ENST00000300403.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002366.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002366.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002366.hg.1 | TC20000198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20002360.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002360.hg.1 | TC20000198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20002363.hg.1 | ENST00000300403.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002363.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002363.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002363.hg.1 | TC20000198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002365.hg.1 | ENST00000300403.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002365.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002365.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002365.hg.1 | TC20000198.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000148367.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148367.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148367.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148367.hg.1 | TC2000007089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148367.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148367.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148367.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148364.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148364.hg.1 | TC2000007089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148364.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148365.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148365.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148365.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148365.hg.1 | TC2000007089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148365.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148365.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148365.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148369.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148369.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148369.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148369.hg.1 | TC2000007089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148369.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148369.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148369.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007089.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148366.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148366.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148366.hg.1 | NM_012112.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148366.hg.1 | TC2000007089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148366.hg.1 | ENST00000300403.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148366.hg.1 | ENST00000340513.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148366.hg.1 | NM_012112.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 189094 | - | - |
