Assay Details
Gene Symbol: MMP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002426.5 | NP_002417.2 | 1 - 2 | 194 | 90 | |
GenBank mRNA | AK313959.1 | - | 1 - 2 | 143 | 90 | |
BC112301.1 | - | 1 - 2 | 194 | 8327769 | 90 | |
BC143773.1 | - | 1 - 2 | 194 | 9052287 | 90 | |
BE440047.1 | - | 1 - 2 | 144 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002237.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
response to organic cyclic compound
extracellular matrix disassembly
collagen catabolic process
wound healing, spreading of epidermal cells
response to drug
positive regulation of epithelial cell proliferation involved in wound healing
endopeptidase activity
metalloendopeptidase activity
serine-type endopeptidase activity
calcium ion binding
zinc ion binding
proteolysis
response to organic cyclic compound
extracellular matrix disassembly
collagen catabolic process
wound healing, spreading of epidermal cells
response to drug
positive regulation of epithelial cell proliferation involved in wound healing
endopeptidase activity
metalloendopeptidase activity
serine-type endopeptidase activity
calcium ion binding
zinc ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11026744.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026744.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026744.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11026744.hg.1 | TC11002237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11026743.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026743.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026743.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11026743.hg.1 | TC11002237.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100193311.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193311.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193311.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193311.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193311.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193311.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193311.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193311.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193311.hg.1 | TC1100012134.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100193309.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193309.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193309.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193309.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193309.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193309.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193309.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193309.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193309.hg.1 | TC1100012134.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100193310.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193310.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193310.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193310.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193310.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193310.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193310.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193310.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193310.hg.1 | TC1100012134.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000326227.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000532855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | ENST00000571244.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012134.hg.1 | NM_002426.5 | Refseq |
