Assay Details
Gene Symbol: CDH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252338.2 | NP_001239267.1 | 3 - 4 | 483 | 105 | |
NM_001252339.2 | NP_001239268.1 | 3 - 4 | 640 | 105 | ||
NM_001794.4 | NP_001785.2 | 4 - 5 | 668 | 105 | ||
GenBank mRNA | AK091496.1 | - | 4 - 5 | 864 | 105 | |
AK124724.1 | - | 3 - 4 | 623 | 105 | ||
AK308345.1 | - | 3 - 4 | 482 | 105 | ||
AK315192.1 | - | 4 - 5 | 622 | 105 | ||
BC101651.1 | - | 4 - 5 | 668 | 8069157 | 105 | |
BC112150.1 | - | 4 - 5 | 668 | 8327618 | 105 | |
L34059.1 | - | 4 - 5 | 594 | 105 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000481.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20006695.hg.1 | ENST00000360469.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20006695.hg.1 | ENST00000536643.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20006695.hg.1 | ENST00000543233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20006695.hg.1 | NM_001252338.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20006695.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20006695.hg.1 | NM_001794.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20006695.hg.1 | TC20000481.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000155478.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155478.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155478.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155478.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000155478.hg.1 | TC2000007973.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000155478.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155478.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155478.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155478.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
