Assay Details
Gene Symbol: CDH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252338.2 | NP_001239267.1 | 9 - 10 | 1528 | 58 | |
NM_001252339.2 | NP_001239268.1 | 9 - 10 | 1685 | 58 | ||
NM_001794.4 | NP_001785.2 | 10 - 11 | 1713 | 58 | ||
GenBank mRNA | AK091496.1 | - | 10 - 11 | 1909 | 58 | |
AK308345.1 | - | 9 - 10 | 1527 | 58 | ||
AK315192.1 | - | 10 - 11 | 1667 | 58 | ||
BC101651.1 | - | 10 - 11 | 1713 | 8069157 | 58 | |
BC112150.1 | - | 10 - 11 | 1713 | 8327618 | 58 | |
L34059.1 | - | 10 - 11 | 1639 | 58 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000481.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20006701.hg.1 | ENST00000360469.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20006701.hg.1 | ENST00000536643.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20006701.hg.1 | ENST00000543233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20006701.hg.1 | NM_001252338.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20006701.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20006701.hg.1 | NM_001794.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20006701.hg.1 | TC20000481.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000155484.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155484.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155484.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155484.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000155484.hg.1 | TC2000007973.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000155484.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155484.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155484.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000155484.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
