Assay Details
| Gene Symbol: MORC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320445.1 | NP_001307374.1 | 13 - 14 | 1620 | 81 | |
| NM_001320446.1 | NP_001307375.1 | 15 - 16 | 1779 | 81 | ||
| NM_015358.2 | NP_056173.1 | 14 - 15 | 1693 | 81 | ||
| XM_011529518.2 | XP_011527820.1 | 14 - 15 | 1913 | 81 | ||
| XM_017028311.1 | XP_016883800.1 | 14 - 15 | 1715 | 81 | ||
| XM_017028312.1 | XP_016883801.1 | 13 - 14 | 1538 | 81 | ||
| GenBank mRNA | AK292957.1 | - | 14 - 15 | 1644 | 81 | |
| AK295133.1 | - | 13 - 14 | 1612 | 81 | ||
| BC007311.1 | - | 14 - 15 | 1650 | 81 | ||
| BC094779.1 | - | 14 - 15 | 1650 | 30527767 | 81 | |
| BC132731.1 | - | 14 - 15 | 1647 | 40146753 | 81 | |
| BC144116.1 | - | 15 - 16 | 1733 | 81 | ||
| D50926.2 | - | 14 - 15 | 1652 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21001062.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21001331.hg.1 | TC21001062.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001331.hg.1 | ENST00000400485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001331.hg.1 | ENST00000487909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001331.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001333.hg.1 | TC21001062.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001333.hg.1 | ENST00000400485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001333.hg.1 | ENST00000487909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001333.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001334.hg.1 | TC21001062.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001334.hg.1 | ENST00000400485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001334.hg.1 | ENST00000487909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001334.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000397184.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000397184.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000397184.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000485299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000485299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000485299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000485299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000485933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000485933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000485933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100158091.hg.1 | TC2100008506.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100158091.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158091.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158091.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100158091.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158091.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158091.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100158090.hg.1 | TC2100008506.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100158090.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158090.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158090.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158090.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100158090.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158090.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158090.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158090.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000397184.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000397184.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000397184.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000400485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000484028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000485299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000485299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000485299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000485299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000485933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000485933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000485933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000487909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | ENST00000492336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008506.hg.1 | NM_015358.2 | Refseq |

MORC3
Genomic Map