Assay Details
| Gene Symbol: LHCGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000233.3 | NP_000224.2 | 9 - 10 | 932 | 143 | |
| XM_011532828.1 | XP_011531130.1 | 8 - 9 | 840 | 143 | ||
| XM_011532831.1 | XP_011531133.1 | 5 - 6 | 563 | 143 | ||
| XM_017004090.1 | XP_016859579.1 | 3 - 4 | 707 | 143 | ||
| GenBank mRNA | M63108.1 | - | 9 - 10 | 870 | 143 | |
| S57793.1 | - | 9 - 10 | 862 | 143 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001830.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
phospholipase C-activating G-protein coupled receptor signaling pathway
male gonad development
hormone-mediated signaling pathway
ovulation cycle process
male genitalia development
positive regulation of inositol trisphosphate biosynthetic process
luteinizing hormone signaling pathway
positive regulation of cAMP-mediated signaling
cognition
cellular response to gonadotropin stimulus
luteinizing hormone receptor activity
G-protein coupled peptide receptor activity
choriogonadotropin hormone receptor activity
choriogonadotropin hormone binding
G-protein coupled receptor signaling pathway
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
phospholipase C-activating G-protein coupled receptor signaling pathway
male gonad development
hormone-mediated signaling pathway
ovulation cycle process
male genitalia development
positive regulation of inositol trisphosphate biosynthetic process
luteinizing hormone signaling pathway
positive regulation of cAMP-mediated signaling
cognition
cellular response to gonadotropin stimulus
luteinizing hormone receptor activity
G-protein coupled peptide receptor activity
choriogonadotropin hormone receptor activity
choriogonadotropin hormone binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029282.hg.1 | TC02001830.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029282.hg.1 | ENST00000294954.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029282.hg.1 | ENST00000344775.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029282.hg.1 | ENST00000401907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029282.hg.1 | ENST00000403273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029282.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226269.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226269.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226269.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226269.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226269.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226269.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226269.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226269.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226269.hg.1 | TC0200016666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |

LHCGR
Genomic Map