Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024432.hg.1 |
TC07001611.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024432.hg.1 |
ENST00000222543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024432.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024433.hg.1 |
TC07001611.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024433.hg.1 |
ENST00000222543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024433.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000222543.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000451238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
ENST00000461482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_001271004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011809.hg.1 |
NM_006528.3 |
Refseq |