Assay Details
| Gene Symbol: PLA2G12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270244.4 | XP_005270301.1 | 2 - 3 | 646 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G12B| Gene Symbol: PLA2G12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270244.4 | XP_005270301.1 | 2 - 3 | 646 | 70 | |
Set Membership: |
|