Assay Details
Gene Symbol: MED18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017638.2 | NP_060108.2 | 1 - 2 | 179 | 103 | |
GenBank mRNA | AB107222.1 | - | 1 - 2 | 135 | 103 | |
AK000052.1 | - | 1 - 2 | 143 | 103 | ||
AK296130.1 | - | 1 - 2 | 149 | 103 | ||
BC002694.2 | - | 1 - 2 | 126 | 3607295 | 103 | |
DA090416.1 | - | 1 - 2 | 211 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000378.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB107222.1,AK000052.1,AK296130.1,BC002694.2,NM_017638.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01005820.hg.1 | TC01000378.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005820.hg.1 | ENST00000479574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005820.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005820.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005824.hg.1 | TC01000378.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005824.hg.1 | ENST00000373842.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005824.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005824.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005823.hg.1 | TC01000378.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005823.hg.1 | ENST00000373842.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005823.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005823.hg.1 | ENST00000479574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005823.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005823.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005826.hg.1 | TC01000378.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005826.hg.1 | ENST00000373842.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005826.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005826.hg.1 | ENST00000479574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005826.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005826.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152858.hg.1 | TC0100007555.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100152855.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152855.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152855.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152855.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152855.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152855.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152855.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152855.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152855.hg.1 | TC0100007555.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152857.hg.1 | TC0100007555.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100152851.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152851.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152851.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152851.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152851.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152851.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152851.hg.1 | TC0100007555.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100152854.hg.1 | TC0100007555.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
