Assay Details
| Gene Symbol: ITGB1BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012278.2 | NP_036410.1 | 3 - 4 | 227 | 72 | |
| GenBank mRNA | AF110225.1 | - | 3 - 4 | 181 | 72 | |
| AF140690.1 | - | 3 - 4 | 197 | 72 | ||
| BC108901.2 | - | 2 - 3 | 454 | 40004692 | 72 | |
| DA895605.1 | - | 3 - 4 | 230 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151992.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151992.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151992.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151992.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151992.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151992.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151992.hg.1 | TC0X00007562.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00151989.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151989.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151989.hg.1 | TC0X00007562.hg.1 | Clariom D TC |

ITGB1BP2
Genomic Map