Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006681.hg.1 |
ENST00000454684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006681.hg.1 |
ENST00000454684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006681.hg.1 |
ENST00000454684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027693.hg.1 |
TC06002210.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027693.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027693.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027694.hg.1 |
TC06002210.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027694.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027692.hg.1 |
TC06002210.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027692.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027692.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
ENST00000340881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013523.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193325.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193325.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193325.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193325.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193325.hg.1 |
TC0600013523.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700006681.hg.1 |
ENST00000454684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006681.hg.1 |
ENST00000454684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006681.hg.1 |
ENST00000454684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193324.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193324.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193324.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193324.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193324.hg.1 |
TC0600013523.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193327.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193327.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193327.hg.1 |
TC0600013523.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193326.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193326.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193326.hg.1 |
ENST00000253329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193326.hg.1 |
NM_139126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193326.hg.1 |
TC0600013523.hg.1 |
Clariom D TC |