Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000474456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000474456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000474456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000474456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039423.hg.1 |
TC01006351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039423.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039423.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241819.hg.1 |
TC0100018433.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000373001.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000474456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000474456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000474456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000474456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000493015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
ENST00000496778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_001271851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018433.hg.1 |
NM_022157.3 |
Refseq |