Assay Details
| Gene Symbol: HIST1H4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003546.2 | NP_003537.1 | 1 | 309 | 63 | |
| GenBank mRNA | BC069392.1 | - | 1 | 309 | 7262681 | 63 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001428.hg.1
Set Membership: |
|

HIST1H4L
Genomic Map