Assay Details
| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005253217.3 | XP_005253274.1 | 3 - 4 | 334 | 104 | |
| XM_006718372.3 | XP_006718435.1 | 3 - 4 | 421 | 104 | ||
| XM_017018530.1 | XP_016874019.1 | 3 - 4 | 367 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005253217.3, XM_006718372.3, XM_017018530.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000340160.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000517621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000517621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000517621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000517621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000517621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000517621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000517621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000517621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000518429.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000518429.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000518429.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000518429.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000518429.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000518429.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000518429.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000518429.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000519051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000519051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000519051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000519051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000519051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000519051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000520999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000521990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000521990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000521990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000521990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000521990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000521990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | ENST00000533786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007400.hg.1 | NM_130783.4 | Refseq |

TSPAN18
Genomic Map