Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005557.hg.1 |
ENST00000430627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005557.hg.1 |
ENST00000447079.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005557.hg.1 |
NM_015083.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005557.hg.1 |
NM_016507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005557.hg.1 |
TC17000474.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
ENST00000430627.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
ENST00000447079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
ENST00000584632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
NM_015083.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
NM_016507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
ENST00000430627.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
ENST00000447079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
ENST00000584632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
NM_015083.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
NM_016507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202066.hg.1 |
TC1700012244.hg.1 |
Clariom D TC |