Assay Details
Gene Symbol: SPG21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127889.4 | NP_001121361.1 | 7 - 8 | 799 | 112 | |
NM_001127890.4 | NP_001121362.1 | 6 - 7 | 899 | 112 | ||
NM_016630.6 | NP_057714.1 | 7 - 8 | 980 | 112 | ||
XM_005254437.4 | XP_005254494.1 | 8 - 9 | 1013 | 112 | ||
XM_017022297.1 | XP_016877786.1 | 7 - 8 | 1348 | 112 | ||
XM_017022298.1 | XP_016877787.1 | 8 - 9 | 1438 | 112 | ||
GenBank mRNA | AF208861.1 | - | 6 - 7 | 695 | 112 | |
AF212231.1 | - | 7 - 8 | 767 | 112 | ||
AK123725.1 | - | 5 - 6 | 1212 | 112 | ||
AK301362.1 | - | 6 - 7 | 864 | 112 | ||
AK312943.1 | - | 7 - 8 | 815 | 112 | ||
BC000244.1 | - | 7 - 8 | 850 | 3357388 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
