Assay Details
| Gene Symbol: CAMKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172214.2 | NP_757363.1 | 16 - 17 | 2425 | 81 | |
| NM_172215.2 | NP_757364.1 | 15 - 16 | 2296 | 81 | ||
| XM_005253823.1 | XP_005253880.1 | 16 - 17 | 1727 | 81 | ||
| XM_017018703.1 | XP_016874192.1 | 17 - 18 | 1781 | 81 | ||
| XM_017018704.1 | XP_016874193.1 | 16 - 17 | 1778 | 81 | ||
| XM_017018708.1 | XP_016874197.1 | 16 - 17 | 1773 | 81 | ||
| GenBank mRNA | AF091074.1 | - | 7 - 8 | 585 | 81 | |
| AF101264.1 | - | 15 - 16 | 1248 | 81 | ||
| AF287631.1 | - | 16 - 17 | 2419 | 81 | ||
| AF321385.1 | - | 15 - 16 | 2046 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002061.hg.1
Set Membership: |
|

CAMKK2
Genomic Map