Assay Details
| Gene Symbol: CAMKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270485.1 | NP_001257414.1 | 3 - 4 | 699 | 117 | |
| NM_001270486.1 | NP_001257415.1 | 2 - 3 | 583 | 117 | ||
| NM_006549.3 | NP_006540.3 | 3 - 4 | 1353 | 117 | ||
| NM_153499.2 | NP_705719.2 | 3 - 4 | 1353 | 117 | ||
| NM_153500.1 | NP_705720.1 | 3 - 4 | 1353 | 117 | ||
| NM_172214.2 | NP_757363.1 | 3 - 4 | 1353 | 117 | ||
| NM_172215.2 | NP_757364.1 | 3 - 4 | 1353 | 117 | ||
| NM_172216.1 | NP_757365.1 | 3 - 4 | 1353 | 117 | ||
| NM_172226.2 | NP_757380.1 | 3 - 4 | 699 | 117 | ||
| XM_005253822.3 | XP_005253879.1 | 3 - 4 | 655 | 117 | ||
| XM_005253823.1 | XP_005253880.1 | 3 - 4 | 655 | 117 | ||
| XM_005253824.3 | XP_005253881.1 | 3 - 4 | 655 | 117 | ||
| XM_011537763.1 | XP_011536065.1 | 3 - 4 | 655 | 117 | ||
| XM_017018698.1 | XP_016874187.1 | 3 - 4 | 706 | 117 | ||
| XM_017018701.1 | XP_016874190.1 | 3 - 4 | 706 | 117 | ||
| XM_017018702.1 | XP_016874191.1 | 3 - 4 | 864 | 117 | ||
| XM_017018704.1 | XP_016874193.1 | 3 - 4 | 706 | 117 | ||
| XM_017018709.1 | XP_016874198.1 | 3 - 4 | 658 | 117 | ||
| GenBank mRNA | AB018330.2 | - | 2 - 3 | 529 | 117 | |
| AB081336.1 | - | 2 - 3 | 556 | 117 | ||
| AB081337.1 | - | 2 - 3 | 556 | 117 | ||
| AF101264.1 | - | 2 - 3 | 176 | 117 | ||
| AF140507.1 | - | 3 - 4 | 637 | 117 | ||
| AF287630.1 | - | 3 - 4 | 1347 | 117 | ||
| AF287631.1 | - | 3 - 4 | 1347 | 117 | ||
| AF321385.1 | - | 3 - 4 | 1103 | 117 | ||
| AF321386.1 | - | 3 - 4 | 1103 | 117 | ||
| AF321387.1 | - | 3 - 4 | 1103 | 117 | ||
| AF321388.1 | - | 3 - 4 | 1103 | 117 | ||
| AK292072.1 | - | 3 - 4 | 847 | 117 | ||
| AK300865.1 | - | 4 - 5 | 599 | 117 | ||
| BC026060.2 | - | 3 - 4 | 632 | 3842706 | 117 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002061.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017018709.1 transcripts.
Set Membership: |
|

CAMKK2
Genomic Map