| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013585.hg.1 |
ENST00000357552.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013585.hg.1 |
ENST00000371001.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013585.hg.1 |
NM_014258.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013585.hg.1 |
TC20000999.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000167471.hg.1 |
ENST00000357552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167471.hg.1 |
ENST00000371001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167471.hg.1 |
ENST00000412613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167471.hg.1 |
NM_014258.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000167471.hg.1 |
TC2000009673.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000167471.hg.1 |
ENST00000357552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167471.hg.1 |
ENST00000371001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167471.hg.1 |
ENST00000412613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167471.hg.1 |
NM_014258.3 |
Refseq |