Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011276.hg.1 |
TC01000703.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011276.hg.1 |
ENST00000371156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011276.hg.1 |
ENST00000371158.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011276.hg.1 |
ENST00000395513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011276.hg.1 |
ENST00000472512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011276.hg.1 |
NM_176877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160667.hg.1 |
ENST00000371158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160667.hg.1 |
ENST00000472512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160667.hg.1 |
ENST00000484937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160667.hg.1 |
NM_176877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160667.hg.1 |
ENST00000371158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160667.hg.1 |
ENST00000472512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160667.hg.1 |
ENST00000484937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160667.hg.1 |
NM_176877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160667.hg.1 |
TC0100008536.hg.1 |
Clariom D TC |