Assay Details
Gene Symbol: NEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000434.3 | NP_000425.1 | 4 - 5 | 950 | 106 | |
GenBank mRNA | AF040958.1 | - | 4 - 5 | 923 | 106 | |
AK290966.1 | - | 4 - 5 | 838 | 106 | ||
AK313006.1 | - | 4 - 5 | 837 | 106 | ||
BC000722.2 | - | 4 - 5 | 891 | 3506824 | 106 | |
BC011900.2 | - | 4 - 5 | 822 | 4101695 | 106 | |
BT007206.1 | - | 4 - 5 | 794 | 106 | ||
CR456717.1 | - | 4 - 5 | 794 | 106 | ||
CR541916.1 | - | 4 - 5 | 794 | 106 | ||
U84246.1 | - | 4 - 5 | 848 | 106 | ||
X78687.1 | - | 4 - 5 | 860 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001548.hg.1, TC6_apd_hap1000089.hg.1, TC6_cox_hap2000175.hg.1, TC6_dbb_hap3000164.hg.1, TC6_mann_hap4000146.hg.1, TC6_mcf_hap5000150.hg.1, TC6_qbl_hap6000165.hg.1, TC6_ssto_hap7000144.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycosphingolipid metabolic process
ganglioside catabolic process
oligosaccharide catabolic process
positive regulation of neuron projection development
regulation of myoblast proliferation
exo-alpha-sialidase activity
exo-alpha-(2->3)-sialidase activity
exo-alpha-(2->6)-sialidase activity
exo-alpha-(2->8)-sialidase activity
glycosphingolipid metabolic process
ganglioside catabolic process
oligosaccharide catabolic process
positive regulation of neuron projection development
regulation of myoblast proliferation
exo-alpha-sialidase activity
exo-alpha-(2->3)-sialidase activity
exo-alpha-(2->6)-sialidase activity
exo-alpha-(2->8)-sialidase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002913.hg.1 | TC6_mcf_hap5000150.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002913.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003149.hg.1 | TC6_qbl_hap6000165.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003149.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06017963.hg.1 | TC06001548.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06017963.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06017963.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003256.hg.1 | TC6_cox_hap2000175.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003256.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003161.hg.1 | TC6_dbb_hap3000164.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003161.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002887.hg.1 | TC6_ssto_hap7000144.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002887.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06017964.hg.1 | TC06001548.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06017964.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06017964.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001314.hg.1 | TC6_apd_hap1000089.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001314.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002888.hg.1 | TC6_ssto_hap7000144.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002888.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003253.hg.1 | TC6_cox_hap2000175.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003253.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003254.hg.1 | TC6_cox_hap2000175.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003254.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003163.hg.1 | TC6_dbb_hap3000164.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003163.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002651.hg.1 | TC6_mann_hap4000146.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002651.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003146.hg.1 | TC6_qbl_hap6000165.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003146.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002890.hg.1 | TC6_ssto_hap7000144.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002890.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001311.hg.1 | TC6_apd_hap1000089.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001311.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002915.hg.1 | TC6_mcf_hap5000150.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002915.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001312.hg.1 | TC6_apd_hap1000089.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001312.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003160.hg.1 | TC6_dbb_hap3000164.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003160.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002650.hg.1 | TC6_mann_hap4000146.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002650.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002653.hg.1 | TC6_mann_hap4000146.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002653.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06017966.hg.1 | TC06001548.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06017966.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06017966.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002912.hg.1 | TC6_mcf_hap5000150.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002912.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003147.hg.1 | TC6_qbl_hap6000165.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003147.hg.1 | NM_000434.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011463.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177737.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177737.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177737.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177737.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177737.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177737.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177737.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177737.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177737.hg.1 | TC0600011463.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600177738.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177738.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177738.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177738.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177738.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177738.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177738.hg.1 | TC0600011463.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600177740.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177740.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177740.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177740.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177740.hg.1 | ENST00000375631.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177740.hg.1 | ENST00000480384.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177740.hg.1 | ENST00000491768.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177740.hg.1 | ENST00000495807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600177740.hg.1 | TC0600011463.hg.1 | Clariom D TC |
