Assay Details
| Gene Symbol: PGAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291730.1 | NP_001278659.1 | 5 - 6 | 611 | 68 | |
| NM_001291732.1 | NP_001278661.1 | 4 - 5 | 548 | 68 | ||
More Information
Set Membership: |
|

PGAP3
Genomic Map