Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016308.hg.1 |
TC06001401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016308.hg.1 |
ENST00000211936.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016308.hg.1 |
ENST00000341087.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016308.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016308.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000554049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000554049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000554049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000555138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000555138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000555138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000585498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000585498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000585816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000585816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000586657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000586657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000586657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000587562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000592921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000592921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000592921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016307.hg.1 |
TC06001401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016307.hg.1 |
ENST00000211936.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016307.hg.1 |
ENST00000341087.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016307.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016307.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008577.hg.1 |
ENST00000527440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008577.hg.1 |
ENST00000527440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008577.hg.1 |
ENST00000527440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008577.hg.1 |
ENST00000527440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175634.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175634.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175634.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175634.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175634.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175634.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175634.hg.1 |
TC0600011197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000554049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000554049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000554049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000555138.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000555138.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000555138.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000555146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010858.hg.1 |
ENST00000556750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175635.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175635.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175635.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175635.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175635.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175635.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175635.hg.1 |
TC0600011197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000585498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000585498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000585816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000585816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000586657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000586657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000586657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000587562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000590606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000592921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000592921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000592921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010099.hg.1 |
ENST00000593655.1 |
Ensembl |