Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016306.hg.1 |
TC06001401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016306.hg.1 |
ENST00000211936.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016306.hg.1 |
ENST00000341087.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016306.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016306.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175633.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175633.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175633.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175633.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175633.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175633.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175633.hg.1 |
TC0600011197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000211936.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
ENST00000377419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011197.hg.1 |
NM_007149.2 |
Refseq |