Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011010.hg.1 |
TC01000686.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011010.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011010.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160094.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160094.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160094.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160094.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160094.hg.1 |
TC0100008454.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
ENST00000361249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008454.hg.1 |
NM_000562.2 |
Refseq |