Assay Details
| Gene Symbol: MRPL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016491.3 | NP_057575.2 | 2 - 3 | 610 | 95 | |
| XM_005270920.3 | XP_005270977.1 | 2 - 3 | 610 | 95 | ||
| GenBank mRNA | AF151069.1 | - | 2 - 3 | 571 | 95 | |
| AF325707.1 | - | 2 - 3 | 558 | 95 | ||
| AK024572.1 | - | 2 - 3 | 610 | 95 | ||
| AK026171.1 | - | 2 - 3 | 594 | 95 | ||
| BC000041.2 | - | 2 - 3 | 555 | 3505543 | 95 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000663.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01010744.hg.1 | TC01000663.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010744.hg.1 | ENST00000329505.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010744.hg.1 | ENST00000336230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010744.hg.1 | ENST00000360840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010744.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010739.hg.1 | TC01000663.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010739.hg.1 | ENST00000360840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010739.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010747.hg.1 | TC01000663.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010747.hg.1 | ENST00000329505.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010747.hg.1 | ENST00000360840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010747.hg.1 | ENST00000487096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010747.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010745.hg.1 | TC01000663.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010745.hg.1 | ENST00000329505.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010745.hg.1 | ENST00000360840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010745.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000398219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000398219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000398219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000398219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010746.hg.1 | TC01000663.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010746.hg.1 | ENST00000329505.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010746.hg.1 | ENST00000360840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010746.hg.1 | ENST00000487096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010746.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159669.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159669.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159669.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159669.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159669.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159669.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159669.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159669.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159669.hg.1 | TC0100008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100159664.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159664.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159664.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159664.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159664.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159664.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159664.hg.1 | TC0100008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159671.hg.1 | TC0100008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000336230.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000398219.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000398219.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000398219.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000398219.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008383.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | ENST00000360840.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | ENST00000487096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | ENST00000490670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | ENST00000605337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | NM_016491.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159670.hg.1 | TC0100008383.hg.1 | Clariom D TC |

MRPL37
Genomic Map