Assay Details
| Gene Symbol: SETD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005253858.4 | XP_005253915.1 | 9 - 10 | 3479 | 64 | |
| XM_006719296.3 | XP_006719359.1 | 9 - 10 | 3395 | 64 | ||
| GenBank mRNA | JF813787.1 | - | 8 - 9 | 3314 | 64 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |

SETD1B
Genomic Map