Assay Details
| Gene Symbol: SETD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015048.1 | NP_055863.1 | 14 - 15 | 5346 | 74 | |
| XM_005253858.4 | XP_005253915.1 | 14 - 15 | 5634 | 74 | ||
| XM_006719296.3 | XP_006719359.1 | 14 - 15 | 5550 | 74 | ||
| GenBank mRNA | AB028999.1 | - | 4 - 5 | 1984 | 74 | |
| JF813787.1 | - | 13 - 14 | 5469 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000963.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12012559.hg.1 | ENST00000267197.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012559.hg.1 | ENST00000542440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012559.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012559.hg.1 | TC12000963.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167967.hg.1 | TC1200009183.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000267197.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000542440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000604567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | ENST00000619791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009183.hg.1 | NM_015048.1 | Refseq |

SETD1B
Genomic Map