Assay Details
| Gene Symbol: ALDOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005257949.2 | XP_005258006.1 | 2 - 3 | 215 | 78 | |
| XM_011524556.1 | XP_011522858.1 | 2 - 3 | 144 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005257949.2, XM_011524556.1 transcripts.
Set Membership: |
|

ALDOC
Genomic Map