Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000397143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000397143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000397143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016370.hg.1 |
TC07001065.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016370.hg.1 |
ENST00000407559.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016370.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000397143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000397143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000397143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013493.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700199600.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199600.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199600.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199600.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700199600.hg.1 |
ENST00000407559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199600.hg.1 |
ENST00000444851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199600.hg.1 |
ENST00000467220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199600.hg.1 |
NM_018051.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700199600.hg.1 |
TC0700013493.hg.1 |
Clariom D TC |