Assay Details
Gene Symbol: WDR60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018051.4 | NP_060521.4 | 2 - 3 | 224 | 59 | |
XM_006716041.2 | XP_006716104.1 | 2 - 3 | 239 | 59 | ||
XM_011516367.1 | XP_011514669.1 | 2 - 3 | 81 | 59 | ||
XM_011516368.2 | XP_011514670.1 | 2 - 3 | 239 | 59 | ||
XM_011516369.2 | XP_011514671.1 | 2 - 3 | 70 | 59 | ||
XM_017012381.1 | XP_016867870.1 | 2 - 3 | 261 | 59 | ||
XM_017012382.1 | XP_016867871.1 | 2 - 3 | 239 | 59 | ||
XM_017012383.1 | XP_016867872.1 | 2 - 3 | 300 | 59 | ||
GenBank mRNA | BC014491.1 | - | 2 - 3 | 224 | 4906683 | 59 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001065.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011516369.2,XM_017012381.1,XM_017012383.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000397143.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000397143.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000397143.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07016347.hg.1 | TC07001065.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07016347.hg.1 | ENST00000407559.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07016347.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000397143.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000397143.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000397143.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000444851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | ENST00000467220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013493.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199568.hg.1 | ENST00000397143.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199568.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199568.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199568.hg.1 | ENST00000397143.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199568.hg.1 | ENST00000407559.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199568.hg.1 | NM_018051.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199568.hg.1 | TC0700013493.hg.1 | Clariom D TC |
