Assay Details
Gene Symbol: OTUD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020205.3 | NP_064590.2 | 1 - 2 | 295 | 125 | |
XM_011509782.1 | XP_011508084.1 | 1 - 2 | 292 | 125 | ||
XM_011509786.1 | XP_011508088.1 | 1 - 2 | 292 | 125 | ||
XM_017001850.1 | XP_016857339.1 | 1 - 2 | 292 | 125 | ||
GenBank mRNA | AK299790.1 | - | 1 - 2 | 281 | 125 | |
BC072681.1 | - | 1 - 2 | 232 | 6170513 | 125 | |
BP321045.1 | - | 1 - 2 | 295 | 125 | ||
BP357326.1 | - | 1 - 2 | 295 | 125 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003198.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK299790.1,BC072681.1,NM_020205.3,XM_011509782.1,XM_011509786.1,XM_017001850.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mucosal immune response
immune response
negative regulation of interleukin-8 production
protein K11-linked deubiquitination
negative regulation of I-kappaB kinase/NF-kappaB signaling
protein K63-linked deubiquitination
protein K48-linked deubiquitination
protein deubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of protein localization to nucleus
DNA binding
thiol-dependent ubiquitin-specific protease activity
protein binding
cysteine-type peptidase activity
zinc ion binding
K63-linked polyubiquitin binding
Lys48-specific deubiquitinase activity
negative regulation of transcription from RNA polymerase II promoter
mucosal immune response
immune response
negative regulation of interleukin-8 production
protein K11-linked deubiquitination
negative regulation of I-kappaB kinase/NF-kappaB signaling
protein K63-linked deubiquitination
protein K48-linked deubiquitination
protein deubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of protein localization to nucleus
DNA binding
thiol-dependent ubiquitin-specific protease activity
protein binding
cysteine-type peptidase activity
zinc ion binding
K63-linked polyubiquitin binding
Lys48-specific deubiquitinase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01049040.hg.1 | TC01003198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01049040.hg.1 | ENST00000369135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049040.hg.1 | ENST00000543330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049040.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01049038.hg.1 | TC01003198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01049038.hg.1 | ENST00000369135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049038.hg.1 | ENST00000543330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049038.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01049044.hg.1 | TC01003198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01049044.hg.1 | ENST00000543330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049044.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01049041.hg.1 | TC01003198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01049041.hg.1 | ENST00000369135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049041.hg.1 | ENST00000543330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049041.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217728.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217728.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217728.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217728.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217728.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217728.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217728.hg.1 | TC0100015717.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100217731.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217731.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217731.hg.1 | TC0100015717.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100217727.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217727.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217727.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217727.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217727.hg.1 | TC0100015717.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100217724.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217724.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217724.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217724.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217724.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217724.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217724.hg.1 | TC0100015717.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100217725.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217725.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217725.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217725.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217725.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100217725.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100217725.hg.1 | TC0100015717.hg.1 | Clariom D TC |
