Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049032.hg.1 |
TC01003198.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049032.hg.1 |
ENST00000369135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049032.hg.1 |
ENST00000479905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049032.hg.1 |
ENST00000543330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049032.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015717.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217718.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217718.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217718.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217718.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217718.hg.1 |
ENST00000417191.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217718.hg.1 |
ENST00000581312.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217718.hg.1 |
ENST00000619871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217718.hg.1 |
NM_020205.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217718.hg.1 |
TC0100015717.hg.1 |
Clariom D TC |