Assay Details
| Gene Symbol: MAPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123067.3 | NP_001116539.1 | 3 - 4 | 549 | 94 | |
| NM_001203251.1 | NP_001190180.1 | 3 - 4 | 549 | 94 | ||
| XM_005257364.4 | XP_005257421.1 | 3 - 4 | 329 | 94 | ||
| XM_005257370.4 | XP_005257427.1 | 3 - 4 | 393 | 94 | ||
| GenBank mRNA | AY730549.1 | - | 2 - 3 | 227 | 94 | |
| BI552187.1 | - | 3 - 4 | 404 | 94 | ||
| DN996935.1 | - | 3 - 4 | 557 | 94 | ||

MAPT
Genomic Map