Assay Details
Gene Symbol: MED18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005245914.4 | XP_005245971.1 | 1 - 2 | 96 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005245914.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000373842.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000398997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000474683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000475655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | ENST00000479574.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_001127350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007555.hg.1 | NM_017638.2 | Refseq |
