Assay Details
Gene Symbol: LINC00158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024027.2 | 2 - 3 | 770 | 141 | ||
GenBank mRNA | AY035383.1 | - | 2 - 3 | 769 | 141 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000332.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR21003775.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21003775.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21003775.hg.1 | TC21000332.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | NR_024027.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | TC2100007784.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000332587.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000439840.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | ENST00000440205.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100152500.hg.1 | NR_024027.2 | Refseq |
