Assay Details
| Gene Symbol: BIVM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017693.3 | NP_060163.2 | 1 - 2 | 471 | 150 | |
| GenBank mRNA | AF411385.1 | - | 1 - 2 | 471 | 150 | |
| AK290658.1 | - | 1 - 2 | 340 | 150 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13001720.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF411385.1,AK290658.1,NM_017693.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000257336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000448849.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000474443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000474443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000474443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000481069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000481069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000481069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000481069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000481069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000490317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000491929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000491929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000491929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000491929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000491929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | ENST00000491929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_001159596.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009993.hg.1 | NM_017693.3 | Refseq |

BIVM
Genomic Map