Assay Details
Gene Symbol: WASF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291965.1 | NP_001278894.1 | 2 - 3 | 217 | 75 | |
NM_006646.5 | NP_006637.2 | 2 - 3 | 217 | 75 | ||
XM_011534889.1 | XP_011533191.1 | 2 - 3 | 228 | 75 | ||
GenBank mRNA | AB020707.2 | - | 1 - 2 | 106 | 75 | |
AF454702.1 | - | 2 - 3 | 170 | 75 | ||
AK289845.1 | - | 2 - 3 | 217 | 75 | ||
BC050283.1 | - | 2 - 3 | 217 | 5300292 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000070.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB020707.2,AF454702.1,AK289845.1,BC050283.1,NM_001291965.1,NM_006646.5,XM_011534889.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13000701.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000701.hg.1 | ENST00000361042.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000701.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000701.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000701.hg.1 | TC13000070.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13000697.hg.1 | ENST00000361042.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000697.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000697.hg.1 | TC13000070.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000698.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000698.hg.1 | ENST00000361042.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000698.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000698.hg.1 | TC13000070.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13000699.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000699.hg.1 | ENST00000361042.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000699.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000699.hg.1 | TC13000070.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145457.hg.1 | TC1300006658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300145452.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145452.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145452.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145452.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145452.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145452.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145452.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145452.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145452.hg.1 | TC1300006658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300145454.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145454.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145454.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145454.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145454.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145454.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145454.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145454.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145454.hg.1 | TC1300006658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300145451.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145451.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145451.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145451.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145451.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145451.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145451.hg.1 | TC1300006658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
