Assay Details
Gene Symbol: WASF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006646.5 | NP_006637.2 | 6 - 7 | 765 | 76 | |
XM_011534889.1 | XP_011533191.1 | 6 - 7 | 776 | 76 | ||
XM_011534890.1 | XP_011533192.1 | 5 - 6 | 706 | 76 | ||
GenBank mRNA | AB020707.2 | - | 5 - 6 | 654 | 76 | |
AB026543.1 | - | 4 - 5 | 540 | 76 | ||
AF134305.1 | - | 3 - 4 | 399 | 76 | ||
AF454702.1 | - | 6 - 7 | 718 | 76 | ||
AK289845.1 | - | 6 - 7 | 765 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000070.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000706.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000706.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000706.hg.1 | TC13000070.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13000705.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000705.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000705.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000705.hg.1 | TC13000070.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13000708.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000708.hg.1 | ENST00000361042.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000708.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000708.hg.1 | TC13000070.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1300145462.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145462.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145462.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145462.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145462.hg.1 | TC1300006658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300145461.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145461.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145461.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145461.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145461.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145461.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145461.hg.1 | TC1300006658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | ENST00000496788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006658.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145464.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145464.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145464.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145464.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145464.hg.1 | ENST00000335327.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145464.hg.1 | ENST00000361042.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145464.hg.1 | NM_001291965.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145464.hg.1 | NM_006646.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145464.hg.1 | TC1300006658.hg.1 | Clariom D TC |
