Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000709.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000709.hg.1 |
ENST00000361042.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000709.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000709.hg.1 |
TC13000070.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
ENST00000496788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006658.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145465.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145465.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145465.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145465.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145465.hg.1 |
ENST00000335327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145465.hg.1 |
ENST00000361042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145465.hg.1 |
NM_001291965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145465.hg.1 |
NM_006646.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145465.hg.1 |
TC1300006658.hg.1 |
Clariom D TC |